Assay Details
| Gene Symbol: Cers2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029789.1 | NP_084065.1 | 4 - 5 | 508 | 77 | |
| GenBank mRNA | AK002626.1 | - | 4 - 5 | 76 | ||
| AK050161.1 | - | 4 - 5 | 489 | 77 | ||
| AK146106.1 | - | 4 - 5 | 487 | 77 | ||
| AK159325.1 | - | 4 - 5 | 487 | 77 | ||
| AY029532.1 | - | 4 - 5 | 508 | 77 | ||
| BC006847.1 | - | 4 - 5 | 480 | 3598473 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cers2
Genomic Map