Assay Details
| Gene Symbol: Ttc39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145948.1 | NP_001139420.1 | 14 - 15 | 1327 | 86 | |
| NM_153392.2 | NP_700441.2 | 14 - 15 | 1336 | 86 | ||
| GenBank mRNA | AK048659.1 | - | 14 - 15 | 1337 | 86 | |
| AK076641.1 | - | 14 - 15 | 1290 | 86 | ||
| AK076720.1 | - | 14 - 15 | 1329 | 86 | ||
| AK076772.1 | - | 14 - 15 | 1291 | 86 | ||
| AK172951.1 | - | 14 - 15 | 1259 | 86 | ||
| BC023883.1 | - | 14 - 15 | 1335 | 5150841 | 86 | |
| BC026660.1 | - | 14 - 15 | 1277 | 4988900 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39a
Genomic Map