Assay Details
| Gene Symbol: Ttc39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145948.1 | NP_001139420.1 | 7 - 8 | 650 | 64 | |
| NM_153392.2 | NP_700441.2 | 7 - 8 | 659 | 64 | ||
| GenBank mRNA | AK048659.1 | - | 7 - 8 | 660 | 64 | |
| AK076641.1 | - | 7 - 8 | 612 | 64 | ||
| AK172951.1 | - | 7 - 8 | 582 | 64 | ||
| BC023883.1 | - | 7 - 8 | 658 | 5150841 | 64 | |
| BC026660.1 | - | 7 - 8 | 600 | 4988900 | 64 | |
More Information
Set Membership: |
|

Ttc39a
Genomic Map