Assay Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198305.2 | NP_938047.1 | 11 - 12 | 1800 | 69 | |
| GenBank mRNA | AK154523.1 | - | 11 - 12 | 1738 | 69 | |
| AK170825.1 | - | 11 - 12 | 1738 | 69 | ||
| AK192039.1 | - | 1 - 2 | 151 | 69 | ||
| AY423764.1 | - | 11 - 12 | 1778 | 69 | ||
| BC139309.1 | - | 11 - 12 | 1741 | 8862331 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019585 | BC048408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019585 | BC058738.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019585 | BC024128.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019585 | AK170825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019585 | AY423764.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128760 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019585 | BC048408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019585 | BC058738.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019585 | BC024128.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019585 | AK170825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019585 | AY423764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P185552 | BC048408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P185552 | BC058738.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P185552 | BC024128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P185552 | AK170825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P185552 | AY423764.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24051406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04539978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04809934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04539969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04539981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04912657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04957687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04912658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04645934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04539964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04883638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24051398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608717 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456253_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435257_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608717 | - | - |
| Mouse Genome 430 2.0 Array | 1435257_at | - | - |
| Mouse Genome 430 2.0 Array | 1435257_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231003_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231003_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400035123.mm.1 | NM_198305.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035123.mm.1 | NM_198305.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035123.mm.1 | TC0400004174.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400035121.mm.1 | NM_198305.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035121.mm.1 | NM_198305.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035121.mm.1 | TC0400004174.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400035120.mm.1 | NM_198305.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035120.mm.1 | NM_198305.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035120.mm.1 | TC0400004174.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400035122.mm.1 | NM_198305.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035122.mm.1 | NM_198305.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035122.mm.1 | NR_106118.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400035122.mm.1 | TC0400004174.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435257_PM_at | - | - |
| Mouse Expression Set 430 | 1435257_at | - | - |
| Mouse Expression Set 430 | 1456253_PM_s_at | - | - |
| Mouse Expression Set 430 | 1456253_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435257_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435257_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1660142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1660142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228857 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228857 | - | - |
| mouse-6 v1.1 (Illumina) | scl0231003.1_140-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0231003.1_140-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2924601 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2924601 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0231003.1_140-S | - | - |

Klhl17
Genomic Map