| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400035134.mm.1 |
NM_198305.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035134.mm.1 |
NM_198305.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035134.mm.1 |
TC0400004174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400035135.mm.1 |
NM_198305.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035135.mm.1 |
NM_198305.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035135.mm.1 |
TC0400004174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400035132.mm.1 |
NM_198305.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035132.mm.1 |
NM_198305.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035132.mm.1 |
TC0400004174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400035133.mm.1 |
NM_198305.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035133.mm.1 |
NM_198305.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035133.mm.1 |
TC0400004174.mm.1 |
MTA TC |