Assay Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198305.2 | NP_938047.1 | 5 - 6 | 926 | 77 | |
| GenBank mRNA | AK049214.1 | - | 5 - 6 | 803 | 77 | |
| AK154523.1 | - | 5 - 6 | 864 | 77 | ||
| AK170825.1 | - | 5 - 6 | 864 | 77 | ||
| AY423764.1 | - | 5 - 6 | 904 | 77 | ||
| BC139309.1 | - | 5 - 6 | 867 | 8862331 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl17
Genomic Map