Assay Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198305.2 | NP_938047.1 | 8 - 9 | 1448 | 68 | |
| GenBank mRNA | AK049214.1 | - | 8 - 9 | 1325 | 68 | |
| AK154523.1 | - | 8 - 9 | 1386 | 68 | ||
| AK170825.1 | - | 8 - 9 | 1386 | 68 | ||
| AY423764.1 | - | 8 - 9 | 1426 | 68 | ||
| BC058738.1 | - | 2 - 3 | 217 | 68 | ||
| BC139309.1 | - | 8 - 9 | 1389 | 8862331 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058738.1 transcripts.
Set Membership: |
|

Klhl17
Genomic Map