Assay Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198305.2 | NP_938047.1 | 9 - 10 | 1537 | 57 | |
| GenBank mRNA | AK154523.1 | - | 9 - 10 | 1475 | 57 | |
| AK170825.1 | - | 9 - 10 | 1475 | 57 | ||
| AY423764.1 | - | 9 - 10 | 1515 | 57 | ||
| BC058738.1 | - | 3 - 4 | 306 | 57 | ||
| BC139309.1 | - | 9 - 10 | 1478 | 8862331 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058738.1 transcripts.
Set Membership: |
|

Klhl17
Genomic Map