Assay Details
| Gene Symbol: Sh3rf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146299.1 | NP_001139771.1 | 2 - 3 | 754 | 97 | |
| GenBank mRNA | AK028153.1 | - | 1 - 2 | 154 | 97 | |
| AK033601.1 | - | 2 - 3 | 798 | 97 | ||
| BC137955.1 | - | 2 - 3 | 707 | 8860974 | 97 | |
| BC145060.1 | - | 2 - 3 | 707 | 9053608 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3rf2
Genomic Map