| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900014997.mm.1 |
NM_018831.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014997.mm.1 |
NM_018831.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014997.mm.1 |
TC1900001705.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014994.mm.1 |
NM_018831.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014994.mm.1 |
NM_018831.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014994.mm.1 |
TC1900001705.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014996.mm.1 |
NM_018831.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014996.mm.1 |
NM_018831.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014996.mm.1 |
TC1900001705.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014999.mm.1 |
NM_018831.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014999.mm.1 |
NM_018831.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014999.mm.1 |
TC1900001705.mm.1 |
MTA TC |