Assay Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025811.3 | NP_080087.1 | 3 - 4 | 1205 | 104 | |
| GenBank mRNA | AK002650.1 | - | 3 - 4 | 790 | 104 | |
| AK033384.1 | - | 3 - 4 | 1206 | 104 | ||
| BC041104.1 | - | 1 - 2 | 174 | 104 | ||
| BC050211.1 | - | 1 - 2 | 61 | 104 | ||
| BC095956.1 | - | 3 - 4 | 1149 | 5330002 | 104 | |
| BC138190.1 | - | 3 - 4 | 1042 | 8861209 | 104 | |
| BC138191.1 | - | 3 - 4 | 1042 | 8861210 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002650.1 transcripts.
Set Membership: |
|

Nhlrc2
Genomic Map