Assay Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025811.3 | NP_080087.1 | 4 - 5 | 1336 | 63 | |
| GenBank mRNA | AK002650.1 | - | 4 - 5 | 921 | 63 | |
| AK033384.1 | - | 4 - 5 | 1337 | 63 | ||
| BC041104.1 | - | 2 - 3 | 305 | 63 | ||
| BC050211.1 | - | 2 - 3 | 192 | 63 | ||
| BC095956.1 | - | 4 - 5 | 1280 | 5330002 | 63 | |
| BC138190.1 | - | 4 - 5 | 1173 | 8861209 | 63 | |
| BC138191.1 | - | 4 - 5 | 1173 | 8861210 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nhlrc2
Genomic Map