Assay Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025811.3 | NP_080087.1 | 7 - 8 | 1792 | 76 | |
| GenBank mRNA | AK002650.1 | - | 7 - 8 | 1377 | 76 | |
| AK004564.1 | - | 7 - 8 | 1439 | 76 | ||
| AK033384.1 | - | 7 - 8 | 1793 | 76 | ||
| BC041104.1 | - | 5 - 6 | 761 | 76 | ||
| BC050211.1 | - | 5 - 6 | 647 | 76 | ||
| BC095956.1 | - | 7 - 8 | 1736 | 5330002 | 76 | |
| BC138190.1 | - | 7 - 8 | 1629 | 8861209 | 76 | |
| BC138191.1 | - | 7 - 8 | 1629 | 8861210 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nhlrc2
Genomic Map