Assay Details
| Gene Symbol: Nhlrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025811.3 | NP_080087.1 | 8 - 9 | 1908 | 93 | |
| GenBank mRNA | AK002650.1 | - | 8 - 9 | 1493 | 93 | |
| AK004564.1 | - | 8 - 9 | 1555 | 93 | ||
| AK033384.1 | - | 8 - 9 | 1909 | 93 | ||
| BC041104.1 | - | 6 - 7 | 877 | 93 | ||
| BC050211.1 | - | 6 - 7 | 763 | 93 | ||
| BC095956.1 | - | 8 - 9 | 1852 | 5330002 | 93 | |
| BC138190.1 | - | 8 - 9 | 1745 | 8861209 | 93 | |
| BC138191.1 | - | 8 - 9 | 1745 | 8861210 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nhlrc2
Genomic Map