| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800003304.mm.1 |
NM_001039218.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003304.mm.1 |
NM_001039218.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003304.mm.1 |
TC1800000449.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800003305.mm.1 |
NM_001039218.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003305.mm.1 |
NM_001039218.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003305.mm.1 |
TC1800000449.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800003299.mm.1 |
NM_001039218.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003299.mm.1 |
NM_001039218.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003299.mm.1 |
TC1800000449.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800003303.mm.1 |
NM_001039218.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003303.mm.1 |
NM_001039218.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800003303.mm.1 |
TC1800000449.mm.1 |
MTA TC |