Assay Details
| Gene Symbol: Spink14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039218.2 | NP_001034307.2 | 5 - 6 | 380 | 99 | |
| GenBank mRNA | AV381433.2 | - | 4 - 5 | 487 | 99 | |
| BC147448.1 | - | 4 - 5 | 332 | 9056090 | 99 | |
| BC147476.1 | - | 4 - 5 | 332 | 9056118 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181201 | AK036889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181201 | AV381433.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181201 | NM_001039218.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551310 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30953 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149893 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181201 | AK036889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181201 | AV381433.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181201 | NM_001039218.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31664874 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149893 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31664842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13740925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13740935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13740920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13740934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14083350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13905870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14083356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14083330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31664893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455430 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455428 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800003307.mm.1 | NM_001039218.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003307.mm.1 | NM_001039218.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003307.mm.1 | TC1800000449.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800003308.mm.1 | NM_001039218.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003308.mm.1 | NM_001039218.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800003308.mm.1 | TC1800000449.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 393998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103170390 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103170390 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235721 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103170390 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103170390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2908807 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2908807 | - | - |

Spink14
Genomic Map