Assay Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029565.3 | NP_083841.4 | 3 - 4 | 493 | 70 | |
| GenBank mRNA | AK003252.1 | - | 3 - 4 | 407 | 70 | |
| AK050162.1 | - | 3 - 4 | 411 | 70 | ||
| AK145644.1 | - | 3 - 4 | 414 | 70 | ||
| AK159761.1 | - | 3 - 4 | 421 | 70 | ||
| AK166772.1 | - | 3 - 4 | 408 | 70 | ||
| AK167058.1 | - | 3 - 4 | 407 | 70 | ||
| AK202788.1 | - | 2 - 3 | 209 | 70 | ||
| AK204157.1 | - | 3 - 4 | 226 | 70 | ||
| AX775905.1 | - | 3 - 4 | 428 | 70 | ||
| BC014732.1 | - | 3 - 4 | 397 | 4235248 | 70 | |
| BC018379.1 | - | 3 - 4 | 415 | 4242583 | 70 | |
| BC045145.1 | - | 3 - 4 | 431 | 6475115 | 70 | |
| BC058273.1 | - | 3 - 4 | 408 | 6494379 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem59
Genomic Map