Assay Details
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285891.1 | NP_001272820.1 | 2 - 3 | 364 | 60 | |
| NM_001285892.1 | NP_001272821.1 | 2 - 3 | 364 | 60 | ||
| NM_019748.3 | NP_062722.1 | 2 - 3 | 364 | 60 | ||
| GenBank mRNA | AB024303.1 | - | 2 - 3 | 252 | 60 | |
| AK010313.1 | - | 2 - 3 | 241 | 60 | ||
| AK011772.1 | - | 2 - 3 | 275 | 60 | ||
| AK087556.1 | - | 2 - 3 | 255 | 60 | ||
| AK090012.1 | - | 2 - 3 | 251 | 60 | ||
| AK154139.1 | - | 2 - 3 | 277 | 60 | ||
| AK159672.1 | - | 2 - 3 | 266 | 60 | ||
| AK162789.1 | - | 2 - 3 | 252 | 60 | ||
| AK188153.1 | - | 1 - 2 | 95 | 60 | ||
| BB838763.1 | - | 2 - 3 | 365 | 60 | ||
| BC068164.1 | - | 2 - 3 | 253 | 30359189 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sae1
Genomic Map