Assay Details
| Gene Symbol: 5830416P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028427.1 | 1 - 2 | 111 | 77 | ||
| GenBank mRNA | AK017935.1 | - | 1 - 2 | 112 | 77 | |
| AK158034.1 | - | 1 - 2 | 99 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

5830416P10Rik
Genomic Map