Assay Details
| Gene Symbol: 5830416P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028427.1 | 3 - 4 | 443 | 70 | ||
| GenBank mRNA | AK047177.1 | - | 3 - 4 | 341 | 69 | |
| AK158034.1 | - | 3 - 4 | 431 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | survival motor neuron domain containing 1 | Smndc1 |
2410004J23Rik, 4933440I19Rik |
-59.6kb | Sense | Chr. 19 - 53379214 - 53391819 on NCBI Build GRCm38 |
| Downstream | uncharacterized LOC73899 | LOC73899 | 8.3kb | Anti-sense | Chr. 19 - 53460066 - 53462731 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

5830416P10Rik
Genomic Map