Assay Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008542.2 | NP_001008542.2 | 1 - 2 | 472 | 119 | |
| GenBank mRNA | AK154538.1 | - | 1 - 2 | 462 | 119 | |
| BC064453.1 | - | 1 - 2 | 457 | 6810025 | 119 | |
| D31824.1 | - | 1 - 2 | 312 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxi1
Genomic Map