Assay Details
| Gene Symbol: Prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029942.1 | NP_084218.1 | 1 - 2 | 94 | 140 | |
| GenBank mRNA | AK021193.1 | - | 1 - 2 | 120 | 140 | |
| BC119251.1 | - | 1 - 2 | 94 | 8734000 | 140 | |
| BC120711.1 | - | 1 - 2 | 94 | 40129634 | 140 | |
| BC145035.1 | - | 1 - 2 | 94 | 9053583 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prelid2
Genomic Map