Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 11 - 12 | 2919 | 79 | |
| GenBank mRNA | AK031388.1 | - | 1 - 2 | 2405 | 79 | |
| AK143600.1 | - | 2920 | 79 | |||
| AK173317.1 | - | 3 - 4 | 1852 | 79 | ||
| BC076633.1 | - | 4 - 5 | 1994 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map