Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 12 - 13 | 3098 | 60 | |
| GenBank mRNA | AK052534.1 | - | 12 - 13 | 3050 | 60 | |
| AK143600.1 | - | 3099 | 60 | |||
| AK173317.1 | - | 4 - 5 | 2031 | 60 | ||
| BC076633.1 | - | 5 - 6 | 2173 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map