Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 13 - 14 | 3248 | 76 | |
| GenBank mRNA | AK052534.1 | - | 13 - 14 | 3200 | 76 | |
| AK143600.1 | - | 3249 | 76 | |||
| AK173317.1 | - | 5 - 6 | 2181 | 76 | ||
| BC037000.1 | - | 1 - 2 | 90 | 76 | ||
| BC076633.1 | - | 6 - 7 | 2323 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc2
Genomic Map