Assay Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033373.2 | NP_001028545.2 | 12 - 13 | 1258 | 67 | |
| GenBank mRNA | AK181908.1 | - | 2 - 3 | 68 | ||
| AK219417.1 | - | 2 - 3 | 180 | 67 | ||
| BC147314.1 | - | 12 - 13 | 1258 | 9056364 | 67 | |
| BC147315.1 | - | 12 - 13 | 1258 | 9056370 | 67 | |
| BC147758.1 | - | 12 - 13 | 1258 | 9087483 | 67 | |
| BC147763.1 | - | 12 - 13 | 1258 | 9087488 | 67 | |
| BC172036.1 | - | 12 - 13 | 1255 | 9054322 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding
protein phosphorylation
phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167092 | BC172036.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167092 | BC147314.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167092 | BC147315.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167092 | BC147758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167092 | BC147763.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665909 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816401 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712396 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235498 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164811 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168559 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006862 | AK042881.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167092 | BC172036.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167092 | BC147314.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167092 | BC147315.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167092 | BC147758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167092 | BC147763.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168559 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04112453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04112386 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00844197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00106123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00789861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00180252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00169580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00246066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01117246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00121094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01117242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00589822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01062493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00180255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00411535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00167959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01062494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00180248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00948169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00530969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00816316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00425534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00885393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01068770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00347264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00425541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00121102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00789865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01068767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00411531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00283542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00167963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01062500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00437590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00759220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00953677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00397732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00545576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00347253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00246069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00180250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00948165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20263074 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10346596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346603 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10346594 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100003667.mm.1 | NM_001033373.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003667.mm.1 | NM_001033373.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003667.mm.1 | TC0100000447.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100003666.mm.1 | NM_001033373.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003666.mm.1 | NM_001033373.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003666.mm.1 | TC0100000447.mm.1 | MTA TC |

Cdk15
Genomic Map