Assay Details
| Gene Symbol: Astn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019514.3 | NP_062387.2 | 5 - 6 | 1428 | 69 | |
| NM_207109.2 | NP_996992.1 | 6 - 7 | 1584 | 69 | ||
| GenBank mRNA | AF116573.1 | - | 3 - 4 | 596 | 69 | |
| AK041160.1 | - | 7 - 8 | 1488 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Astn2
Genomic Map