Assay Details
| Gene Symbol: Astn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019514.3 | NP_062387.2 | 13 - 14 | 2525 | 57 | |
| NM_207109.2 | NP_996992.1 | 14 - 15 | 2681 | 57 | ||
| GenBank mRNA | AK041160.1 | - | 15 - 16 | 2585 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Astn2
Genomic Map