Assay Details
| Gene Symbol: Astn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019514.3 | NP_062387.2 | 2 - 3 | 789 | 68 | |
| NM_207109.2 | NP_996992.1 | 2 - 3 | 789 | 68 | ||
| GenBank mRNA | AK041160.1 | - | 3 - 4 | 693 | 68 | |
| AK047058.1 | - | 2 - 3 | 778 | 68 | ||
| AK051472.1 | - | 2 - 3 | 790 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041160.1 transcripts.
Set Membership: |
|

Astn2
Genomic Map