| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400006379.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006379.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006379.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006375.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006375.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006375.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006387.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006387.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006387.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006387.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006387.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006391.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006391.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006391.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006391.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006391.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006382.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006382.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006382.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006382.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006382.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006383.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006383.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006383.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006383.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006383.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006389.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006389.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006389.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006389.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006389.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006373.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006373.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006373.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006385.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006385.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006385.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006385.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006385.mm.1 |
TC0400000961.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006390.mm.1 |
NM_001113412.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006390.mm.1 |
NM_001113412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006390.mm.1 |
NM_029347.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006390.mm.1 |
NM_029347.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006390.mm.1 |
TC0400000961.mm.1 |
MTA TC |