| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
NM_001039157.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
NM_001039158.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
NM_001039159.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
NM_001284312.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
NM_021402.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
NR_002889.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017665.mm.1 |
TC0400002100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
NM_001039157.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
NM_001039158.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
NM_001039159.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
NM_001284312.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
NM_021402.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
NR_002889.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017666.mm.1 |
TC0400002100.mm.1 |
MTA TC |