Assay Details
| Gene Symbol: Mrpl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025570.2 | NP_079846.1 | 2 - 3 | 239 | 91 | |
| GenBank mRNA | AB049645.1 | - | 2 - 3 | 212 | 91 | |
| AK004053.1 | - | 2 - 3 | 224 | 91 | ||
| AK011341.1 | - | 2 - 3 | 240 | 91 | ||
| AK015199.1 | - | 223 | 91 | |||
| AK168665.1 | - | 2 - 3 | 213 | 91 | ||
| BC016591.1 | - | 2 - 3 | 202 | 3158162 | 91 | |
| BC039985.1 | - | 2 - 3 | 209 | 1244957 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl20
Genomic Map