Assay Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007859.4 | NP_031885.3 | 6 - 7 | 875 | 62 | |
| GenBank mRNA | AB009377.1 | - | 6 - 7 | 783 | 62 | |
| AK077619.1 | - | 6 - 7 | 921 | 62 | ||
| AK089267.1 | - | 6 - 7 | 853 | 62 | ||
| BC053052.1 | - | 6 - 7 | 805 | 6403143 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic DNA fragmentation
apoptotic process
apoptotic chromosome condensation
nucleic acid phosphodiester bond hydrolysis
DNA binding
GPI-anchor transamidase activity
nuclease activity
deoxyribonuclease activity
protein binding
hydrolase activity
enzyme binding
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
apoptotic DNA fragmentation
apoptotic process
apoptotic chromosome condensation
nucleic acid phosphodiester bond hydrolysis
DNA binding
GPI-anchor transamidase activity
nuclease activity
deoxyribonuclease activity
protein binding
hydrolase activity
enzyme binding
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428163 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428166 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26777 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156581 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105853 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23428159 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105853 | - | - |
| Whole Mouse Genome G4122A | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | AK089267.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | BC053052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | NM_007859.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P46742 | AK077619.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135230 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105853 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03766314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03766319 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03766330 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03766312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428163 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23428170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23428135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03348037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03915627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03766322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03261781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03675398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03261785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03348040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03261773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03915628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03348034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03261788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23428093 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518966 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518965 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518963 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421229_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437051_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518957 | - | - |
| Mouse Genome 430 2.0 Array | 1437051_at | - | - |
| Mouse Genome 430 2.0 Array | 1421229_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421229_at | - | - |
| Mouse Genome 430 2.0 Array | 1437051_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13368_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13368_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400034241.mm.1 | NM_007859.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034241.mm.1 | NM_007859.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034241.mm.1 | TC0400004110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400034242.mm.1 | NM_007859.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034242.mm.1 | NM_007859.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400034242.mm.1 | TC0400004110.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421229_PM_at | - | - |
| Mouse Expression Set 430 | 1437051_at | - | - |
| Mouse Expression Set 430 | 1437051_PM_at | - | - |
| Mouse Expression Set 430 | 1421229_at | - | - |
| Murine Genome U74v2 Set | 100294_PM_at | - | - |
| Murine Genome U74v2 Set | 100294_at | - | - |
| Murine Genome U74v2 Set | 166801_PM_at | - | - |
| Murine Genome U74v2 Set | 166801_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437051_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437051_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421229_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421229_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421229_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437051_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421229_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437051_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 541970 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1770092 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1770092 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770092 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2608441 | - | - |
| mouse-6 v1.1 (Illumina) | scl23457.9.1_86-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770092 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23457.9.1_86-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770092 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2608441 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608441 | - | - |

Dffb
Genomic Map