Assay Details
| Gene Symbol: Dcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027490.1 | NP_081766.1 | 1 - 2 | 190 | 72 | |
| GenBank mRNA | AK008609.1 | - | 1 - 2 | 188 | 72 | |
| AK010490.1 | - | 1 - 2 | 163 | 72 | ||
| AK017809.1 | - | 1 - 2 | 182 | 72 | ||
| AK153105.1 | - | 1 - 2 | 192 | 72 | ||
| AK166560.1 | - | 1 - 2 | 169 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcp2
Genomic Map