Assay Details
| Gene Symbol: Dcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027490.1 | NP_081766.1 | 7 - 8 | 937 | 122 | |
| GenBank mRNA | AK017809.1 | - | 7 - 8 | 929 | 122 | |
| AK152160.1 | - | 7 - 8 | 910 | 122 | ||
| AK153105.1 | - | 7 - 8 | 939 | 122 | ||
| AK153264.1 | - | 7 - 8 | 910 | 122 | ||
| AK153310.1 | - | 7 - 8 | 910 | 122 | ||
| AK166560.1 | - | 7 - 8 | 916 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcp2
Genomic Map