Assay Details
| Gene Symbol: Dcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027490.1 | NP_081766.1 | 9 - 10 | 1185 | 55 | |
| GenBank mRNA | AK017809.1 | - | 9 - 10 | 1177 | 55 | |
| AK152160.1 | - | 9 - 10 | 1158 | 55 | ||
| AK153105.1 | - | 9 - 10 | 1187 | 55 | ||
| AK153264.1 | - | 9 - 10 | 1158 | 55 | ||
| AK153310.1 | - | 9 - 10 | 1158 | 55 | ||
| AK166560.1 | - | 9 - 10 | 1164 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcp2
Genomic Map