Assay Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147221.2 | NP_671754.2 | 1 - 2 | 177 | 88 | |
| GenBank mRNA | AF486579.1 | - | 1 - 2 | 177 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF486579.1, NM_147221.2 transcripts.
Set Membership: |
|

Glis1
Genomic Map