Assay Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147221.2 | NP_671754.2 | 5 - 6 | 1689 | 88 | |
| GenBank mRNA | AF220434.1 | - | 3 - 4 | 1543 | 88 | |
| AF486579.1 | - | 5 - 6 | 1688 | 88 | ||
| AK145342.1 | - | 2 - 3 | 213 | 88 | ||
| BC066157.1 | - | 3 - 4 | 1124 | 30468539 | 88 | |
| JQ043365.1 | - | 4 - 5 | 1468 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glis1
Genomic Map