Assay Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147221.2 | NP_671754.2 | 9 - 10 | 2267 | 85 | |
| GenBank mRNA | AF220434.1 | - | 7 - 8 | 2121 | 85 | |
| AF486579.1 | - | 9 - 10 | 2266 | 85 | ||
| AK145342.1 | - | 6 - 7 | 792 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glis1
Genomic Map