Assay Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009949.2 | NP_034079.2 | 2 - 3 | 352 | 73 | |
| GenBank mRNA | AK018752.1 | - | 2 - 3 | 301 | 73 | |
| AK144441.1 | - | 2 - 3 | 352 | 73 | ||
| AK169038.1 | - | 2 - 3 | 249 | 73 | ||
| BC138514.1 | - | 2 - 3 | 287 | 8861534 | 73 | |
| BC145859.1 | - | 2 - 3 | 287 | 40131162 | 73 | |
| CD545813.1 | - | 2 - 3 | 327 | 73 | ||
| U01163.1 | - | 2 - 3 | 297 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpt2
Genomic Map