| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
NM_001113559.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
NM_001113559.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
NM_001243163.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
NM_001243163.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
NM_011444.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
NM_011444.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026802.mm.1 |
TC0600003397.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
NM_001113559.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
NM_001113559.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
NM_001243163.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
NM_001243163.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
NM_011444.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
NM_011444.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026800.mm.1 |
TC0600003397.mm.1 |
MTA TC |