Assay Details
| Gene Symbol: Psma8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163609.1 | NP_001157081.1 | 4 - 5 | 560 | 85 | |
| GenBank mRNA | AK010717.1 | - | 4 - 5 | 561 | 85 | |
| BC116990.1 | - | 4 - 5 | 522 | 40126309 | 85 | |
| BC116992.1 | - | 4 - 5 | 522 | 40126311 | 85 | |
| BC144771.1 | - | 5 - 6 | 593 | 9053319 | 85 | |
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC1800000123.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC1800000123.mm.1| Set Membership: |  Amplicon length between 71 and 85  Protease  Mammalian Gene Collection  Recommended Assay  Mus musculus  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Psma8
Psma8 Genomic Map
Genomic Map Back To Top
 Back To Top