| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600026542.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026542.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026542.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026539.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026539.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026539.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026536.mm.1 |
NM_001204906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026536.mm.1 |
NM_001204906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026536.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026543.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026543.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026543.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026541.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026541.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026541.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026535.mm.1 |
NM_001204906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026535.mm.1 |
NM_001204906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026535.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
NM_001204906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
NM_001204906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
NM_001204907.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
NM_001204907.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026550.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026540.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026540.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026540.mm.1 |
TC0600003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
NM_001204906.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
NM_001204906.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
NM_001204907.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
NM_001204907.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
NM_023042.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
NM_023042.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026549.mm.1 |
TC0600003373.mm.1 |
MTA TC |