Assay Details
| Gene Symbol: Axin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015732.4 | NP_056547.3 | 5 - 6 | 1549 | 74 | |
| GenBank mRNA | AF073788.1 | - | 5 - 6 | 1414 | 74 | |
| AF205889.1 | - | 4 - 5 | 1254 | 74 | ||
| AK049192.1 | - | 5 - 6 | 1493 | 74 | ||
| AK142333.1 | - | 5 - 6 | 1551 | 74 | ||
| BC057338.1 | - | 5 - 6 | 1532 | 6827741 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Axin2
Genomic Map