Assay Details
| Gene Symbol: Nfatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164109.1 | NP_001157581.1 | 5 - 6 | 2074 | 79 | |
| NM_001164110.1 | NP_001157582.1 | 5 - 6 | 2074 | 79 | ||
| NM_001164111.1 | NP_001157583.1 | 5 - 6 | 1782 | 79 | ||
| NM_001164112.1 | NP_001157584.1 | 5 - 6 | 1782 | 79 | ||
| NM_016791.4 | NP_058071.2 | 5 - 6 | 1782 | 79 | ||
| NM_198429.2 | NP_940821.1 | 5 - 6 | 2074 | 79 | ||
| GenBank mRNA | AF049606.1 | - | 5 - 6 | 1735 | 79 | |
| AF087434.1 | - | 5 - 6 | 1769 | 79 | ||
| AF239169.1 | - | 5 - 6 | 1772 | 79 | ||
| AK004810.1 | - | 5 - 6 | 1740 | 79 | ||
| AK048610.1 | - | 5 - 6 | 2076 | 79 | ||
| AK144965.1 | - | 5 - 6 | 2075 | 79 | ||
| BC061509.1 | - | 5 - 6 | 2061 | 5354603 | 79 | |
| EU887567.1 | - | 5 - 6 | 1769 | 79 | ||
| EU887568.1 | - | 5 - 6 | 1769 | 79 | ||
| EU887569.1 | - | 5 - 6 | 1727 | 79 | ||
| EU887570.1 | - | 5 - 6 | 1769 | 79 | ||
| EU887571.1 | - | 5 - 6 | 1727 | 79 | ||
| EU887572.1 | - | 5 - 6 | 1727 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc1
Genomic Map