Assay Details
| Gene Symbol: Eif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010123.3 | NP_034253.3 | 19 - 20 | 3585 | 84 | |
| GenBank mRNA | U14172.1 | - | 19 - 20 | 3535 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3a
Genomic Map