Assay Details
| Gene Symbol: Hes5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010419.4 | NP_034549.1 | 1 - 2 | 129 | 73 | |
| GenBank mRNA | AK078170.1 | - | 1 - 2 | 73 | ||
| AK087508.1 | - | 1 - 2 | 130 | 73 | ||
| AK139390.1 | - | 1 - 2 | 131 | 73 | ||
| BC099863.1 | - | 1 - 2 | 81 | 40039952 | 73 | |
| BC099864.1 | - | 1 - 2 | 81 | 40039951 | 73 | |
| BC099865.1 | - | 1 - 2 | 81 | 40039949 | 73 | |
| BC103539.1 | - | 1 - 2 | 79 | 40039948 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hes5
Genomic Map