Assay Details
| Gene Symbol: Slco1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267707.1 | NP_001254636.1 | 1 - 2 | 126 | 122 | |
| GenBank mRNA | AK166446.1 | - | 1 - 2 | 103 | 122 | |
| BC013594.1 | - | 1 - 2 | 88 | 3586426 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC013594.1, NM_001267707.1 transcripts.
Set Membership: |
|

Slco1a5
Genomic Map