Assay Details
| Gene Symbol: Kcnj8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008428.4 | NP_032454.1 | 1 - 2 | 165 | 85 | |
| GenBank mRNA | AK031777.1 | - | 1 - 2 | 181 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_008428.4 transcripts.
Set Membership: |
|

Kcnj8
Genomic Map