Assay Details
| Gene Symbol: Mospd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045438.1 | 1 | 774 | 75 | ||
| GenBank mRNA | AK008233.1 | - | 1 | 776 | 75 | |
| AK076850.1 | - | 1 | 716 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mospd4
Genomic Map